Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267396.3 | NP_001254325.1 | 1 - 2 | 156 | 87 | |
GenBank mRNA | U81043.1 | - | 1 - 2 | 162 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |