Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065063.6 | NP_497464.1 | 3 - 4 | 278 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |