Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381780.2 | NP_001368042.1 | 11 - 12 | 2106 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |