Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065175.7 | NP_497576.1 | 10 - 11 | 2885 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |