Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027473.4 | NP_001022644.1 | 11 - 12 | 2132 | 88 | |
NM_001027475.3 | NP_001022646.1 | 11 - 12 | 1903 | 88 | ||
NM_001267984.4 | NP_001254913.1 | 3 - 4 | 379 | 88 | ||
NM_001384070.1 | 10 - 11 | 1853 | 88 | |||
GenBank mRNA | AF031838.1 | - | 10 - 11 | 1863 | 88 | |
AF031839.1 | - | 10 - 11 | 1860 | 88 | ||
AF031840.1 | - | 11 - 12 | 1910 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |