Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027475.3 | NP_001022646.1 | 8 - 9 | 1533 | 60 | |
GenBank mRNA | AF031840.1 | - | 8 - 9 | 1540 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |