Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047405.6 | NP_001040870.1 | 5 - 6 | 529 | 88 | |
NM_001379736.1 | NP_001367783.1 | 6 - 7 | 624 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |