Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027821.6 | NP_001022992.1 | 11 - 12 | 3274 | 81 | |
NM_065910.7 | NP_498311.2 | 11 - 12 | 3259 | 81 | ||
GenBank mRNA | AF469173.1 | - | 11 - 12 | 3212 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |