Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027528.5 | NP_001022699.1 | 2 - 3 | 293 | 106 | |
NM_001027529.6 | NP_001022700.1 | 2 - 3 | 296 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |