Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066605.8 | NP_499006.1 | 2 - 3 | 232 | 106 | |
GenBank mRNA | X13689.1 | - | 194 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |