Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emb-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066673.7 | NP_499074.2 | 13 - 14 | 2647 | 92 | |
GenBank mRNA | AF192400.1 | - | 13 - 14 | 2566 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |