Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066730.8 | NP_499131.1 | 1 - 2 | 97 | 113 | |
GenBank mRNA | AF287259.1 | - | 1 - 2 | 119 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |