Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268143.3 | NP_001255072.1 | 16 - 17 | 2592 | 90 | |
NM_001268144.3 | NP_001255073.1 | 12 - 13 | 1793 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |