Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nft-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067155.8 | NP_499556.1 | 4 - 5 | 457 | 75 | |
GenBank mRNA | AF069986.1 | - | 4 - 5 | 455 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |