Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nob-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383007.1 | 1 - 2 | 239 | 81 | ||
NR_147825.1 | 1 - 2 | 185 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |