Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: toca-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067437.6 | NP_499838.2 | 9 - 10 | 1206 | 103 | |
NM_067438.5 | NP_499839.2 | 9 - 10 | 1254 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |