Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: toca-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067437.6 | NP_499838.2 | 12 - 13 | 1520 | 86 | |
NM_067438.5 | NP_499839.2 | 12 - 13 | 1568 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |