Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307363.3 | NP_001294292.1 | 11 - 12 | 1135 | 118 | |
NM_001307364.3 | NP_001294293.1 | 10 - 11 | 1141 | 118 | ||
NM_001307365.3 | NP_001294294.1 | 9 - 10 | 1018 | 118 | ||
NM_068737.2 | NP_501138.1 | 11 - 12 | 1138 | 118 | ||
GenBank mRNA | U76402.1 | - | 11 - 12 | 1138 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |