Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307276.3 | NP_001294205.1 | 3 - 4 | 833 | 124 | |
NM_001383136.1 | 7 - 8 | 1427 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |