Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069144.6 | NP_501545.1 | 9 - 10 | 2712 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |