Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028425.5 | NP_001023596.1 | 8 - 9 | 1605 | 117 | |
NM_001028426.5 | NP_001023597.1 | 8 - 9 | 1611 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |