Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383170.1 | 4 - 5 | 1429 | 66 | ||
NM_069369.5 | NP_501770.1 | 5 - 6 | 764 | 66 | ||
NM_069370.4 | NP_501771.1 | 4 - 5 | 494 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |