Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383170.1 | 6 - 7 | 1997 | 95 | ||
NM_069369.5 | NP_501770.1 | 7 - 8 | 1332 | 95 | ||
NM_069370.4 | NP_501771.1 | 6 - 7 | 1062 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |