Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dur-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027974.4 | NP_001023145.1 | 9 - 10 | 596 | 66 | |
NM_001268504.3 | NP_001255433.1 | 12 - 13 | 791 | 66 | ||
NM_001268505.3 | NP_001255434.1 | 13 - 14 | 857 | 66 | ||
NM_001268506.3 | NP_001255435.1 | 13 - 14 | 860 | 66 | ||
NM_001268507.3 | NP_001255436.1 | 15 - 16 | 1694 | 66 | ||
NM_001268510.3 | NP_001255439.1 | 6 - 7 | 374 | 66 | ||
NM_001268514.3 | NP_001255443.1 | 2 - 3 | 131 | 66 | ||
NM_001307013.3 | NP_001293942.1 | 9 - 10 | 527 | 66 | ||
NM_001372910.1 | NP_001360167.1 | 9 - 10 | 596 | 66 | ||
NM_001380423.1 | NP_001368122.1 | 2 - 3 | 131 | 66 | ||
NM_001383174.1 | 9 - 10 | 597 | 66 | |||
NM_001383175.1 | 9 - 10 | 596 | 66 | |||
NM_069385.6 | NP_501786.2 | 13 - 14 | 1572 | 66 | ||
NM_069386.8 | NP_501787.3 | 14 - 15 | 1625 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |