Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pyp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027902.4 | NP_001023073.1 | 2 - 3 | 223 | 79 | |
NM_001027904.7 | NP_001023075.2 | 3 - 4 | 490 | 79 | ||
NM_001027905.7 | NP_001023076.1 | 1 - 2 | 85 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |