Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069537.7 | NP_501938.1 | 2 - 3 | 396 | 122 | |
GenBank mRNA | AF038576.1 | - | 2 - 3 | 417 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |