Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: miz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268583.2 | NP_001255512.1 | 7 - 8 | 1133 | 142 | |
NM_001268584.2 | NP_001255513.1 | 3 - 4 | 611 | 142 | ||
NM_001307057.4 | NP_001293986.1 | 7 - 8 | 1135 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |