Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srt-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067575.3 | NP_499976.3 | 3 - 4 | 602 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |