Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307519.4 | NP_001294448.1 | 3 - 4 | 932 | 75 | |
NM_067742.7 | NP_500143.1 | 3 - 4 | 923 | 75 | ||
GenBank mRNA | AF353302.1 | - | 3 - 4 | 897 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |