Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hlh-30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028240.6 | NP_001023411.1 | 7 - 8 | 931 | 73 | |
NM_001028242.6 | NP_001023413.1 | 5 - 6 | 635 | 73 | ||
NM_001028243.3 | NP_001023414.1 | 5 - 6 | 800 | 73 | ||
NM_001364041.3 | NP_001350991.1 | 6 - 7 | 662 | 73 | ||
NM_001372787.3 | NP_001360026.1 | 6 - 7 | 875 | 73 | ||
NM_001380173.1 | NP_001368701.1 | 7 - 8 | 737 | 73 | ||
NM_001383072.1 | 7 - 8 | 900 | 73 | |||
NM_001383073.1 | 7 - 8 | 871 | 73 | |||
NM_068060.5 | NP_500461.1 | 6 - 7 | 856 | 73 | ||
NM_068061.6 | NP_500462.1 | 6 - 7 | 873 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |