Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380207.2 | NP_001367921.1 | 1 - 2 | 93 | 82 | |
GenBank mRNA | AF502568.1 | - | 2 - 3 | 201 | 82 | |
AF519108.1 | - | 1 - 2 | 82 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |