Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srw-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356920.2 | NP_001343673.1 | 4 - 5 | 717 | 86 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Probe spans exons |