Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306908.3 | NP_001293837.1 | 1 - 2 | 147 | 110 | |
NM_068407.5 | NP_500808.1 | 1 - 2 | 132 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |