Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306820.3 | NP_001293749.1 | 4 - 5 | 513 | 67 | |
NM_001383098.1 | 3 - 4 | 434 | 67 | |||
NM_068421.6 | NP_500822.2 | 3 - 4 | 429 | 67 | ||
NM_068422.6 | NP_500823.1 | 3 - 4 | 431 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |