Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028178.4 | NP_001023349.1 | 6 - 7 | 1316 | 106 | |
NM_001129324.3 | NP_001122796.1 | 6 - 7 | 1316 | 106 | ||
NM_001372925.2 | NP_001360493.1 | 6 - 7 | 1322 | 106 |
Gene Symbol: T01G1.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028180.7 | NP_001023351.1 | 12 - 13 | 2634 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |