Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sms-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136389.4 | NP_001129861.2 | 4 - 5 | 872 | 84 | |
NM_001268630.3 | NP_001255559.1 | 4 - 5 | 1061 | 84 | ||
NM_069767.5 | NP_502168.2 | 7 - 8 | 1293 | 84 | ||
NM_069768.6 | NP_502169.2 | 5 - 6 | 1194 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |