Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 12 - 13 | 1779 | 135 | ||
NM_070018.6 | NP_502419.2 | 7 - 8 | 1047 | 135 | ||
GenBank mRNA | AB055112.1 | - | 9 - 10 | 1444 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |