Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cho-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070138.8 | NP_502539.1 | 3 - 4 | 462 | 73 | |
GenBank mRNA | AB030946.1 | - | 3 - 4 | 463 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |