Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mau-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268805.3 | NP_001255734.1 | 4 - 5 | 719 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |