Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qui-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307155.3 | NP_001294084.1 | 10 - 11 | 2950 | 139 | |
NM_070212.4 | NP_502613.2 | 15 - 16 | 3757 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |