Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269027.3 | NP_001255956.2 | 7 - 8 | 851 | 94 | |
NM_001269028.2 | NP_001255957.2 | 6 - 7 | 881 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |