Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tat-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071457.2 | NP_503858.1 | 4 - 5 | 1116 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() |