Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srx-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071620.3 | NP_504021.1 | 6 - 7 | 939 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |