Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp-35B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071693.3 | NP_504094.1 | 2 - 3 | 377 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |