Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpg-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_071863.7 | NP_504264.1 | 1 - 2 | 196 | 59 | |
GenBank mRNA | DQ340630.1 | - | 1 - 2 | 174 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |