Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nex-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322553.3 | NP_001309625.1 | 4 - 5 | 507 | 98 | |
NM_001380638.1 | NP_001368593.1 | 4 - 5 | 549 | 98 | ||
NM_001383302.1 | 4 - 5 | 541 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |