Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srg-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072023.4 | NP_504424.3 | 5 - 6 | 953 | 174 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |