Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307815.4 | NP_001294744.1 | 5 - 6 | 429 | 101 | |
NM_001356646.3 | NP_001343626.1 | 5 - 6 | 439 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |