Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dbl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072308.8 | NP_504709.1 | 6 - 7 | 1127 | 110 | |
GenBank mRNA | AF004395.1 | - | 6 - 7 | 1085 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |