Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_072370.4 | NP_504771.1 | 6 - 7 | 821 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |